Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127492.1 | NP_001120964.1 | 4 - 5 | 463 | 71 | |
XM_006245230.2 | XP_006245292.1 | 4 - 5 | 635 | 71 | ||
XM_006245231.2 | XP_006245293.1 | 4 - 5 | 1036 | 71 | ||
XM_006245233.2 | XP_006245295.1 | 3 - 4 | 555 | 71 | ||
XM_006245234.2 | XP_006245296.1 | 4 - 5 | 634 | 71 | ||
XM_008767251.1 | XP_008765473.1 | 3 - 4 | 957 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |