Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135596.1 | NP_001129068.1 | 4 - 5 | 779 | 65 | |
GenBank mRNA | BC099202.1 | - | 4 - 5 | 779 | 7384071 | 65 |
FQ211754.1 | - | 5 - 6 | 622 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |