Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135596.1 | NP_001129068.1 | 8 - 9 | 1448 | 87 | |
GenBank mRNA | BC099202.1 | - | 8 - 9 | 1448 | 7384071 | 87 |
FQ211754.1 | - | 8 - 9 | 1171 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |