Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135596.1 | NP_001129068.1 | 10 - 11 | 1621 | 93 | |
GenBank mRNA | BC099202.1 | - | 10 - 11 | 1621 | 7384071 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |