Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106921.1 | NP_001100391.1 | 10 - 11 | 1290 | 64 | |
XM_008767222.1 | XP_008765444.1 | 10 - 11 | 1321 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus Inventoried Probe spans exons |