Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030025.1 | NP_001025196.1 | 2 - 3 | 140 | 89 | |
XM_006251483.2 | XP_006251545.1 | 3 - 4 | 509 | 89 | ||
XM_006251484.2 | XP_006251546.1 | 3 - 4 | 415 | 89 | ||
XM_006251485.2 | XP_006251547.1 | 3 - 4 | 359 | 89 | ||
XM_008770270.1 | XP_008768492.1 | 3 - 4 | 333 | 89 | ||
GenBank mRNA | BC099752.1 | - | 2 - 3 | 140 | 5623209 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |