Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143893.1 | NP_001137365.1 | 1 - 2 | 53 | 88 | |
XM_006254426.2 | XP_006254488.1 | 1 - 2 | 2106 | 88 | ||
XM_008771955.1 | XP_008770177.1 | 1 - 2 | 2106 | 88 | ||
GenBank mRNA | BC160837.1 | - | 1 - 2 | 37 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |