Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ss18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100900.1 | NP_001094370.1 | 5 - 6 | 666 | 76 | |
XM_006254437.2 | XP_006254499.1 | 5 - 6 | 669 | 76 | ||
XM_006254438.2 | XP_006254500.1 | 5 - 6 | 690 | 76 | ||
GenBank mRNA | BC083824.1 | - | 5 - 6 | 646 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |