Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impact | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012235.1 | NP_001012235.1 | 7 - 8 | 669 | 94 | |
GenBank mRNA | AY574213.1 | - | 7 - 8 | 669 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |