Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impact | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012235.1 | NP_001012235.1 | 8 - 9 | 749 | 63 | |
GenBank mRNA | AY574213.1 | - | 8 - 9 | 749 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |