Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100516.1 | NP_001093986.1 | 3 - 4 | 2122 | 123 | |
GenBank mRNA | CK473534.1 | - | 2 - 3 | 222 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |