Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228117.1 | XP_006228179.1 | 1 - 2 | 515 | 66 | |
XM_006228118.1 | XP_006228180.1 | 1 - 2 | 515 | 66 | ||
GenBank mRNA | FQ222002.1 | - | 1 - 2 | 91 | 66 | |
FQ229914.1 | - | 1 - 2 | 33 | 66 | ||
FQ230074.1 | - | 1 - 2 | 126 | 66 | ||
FQ233974.1 | - | 1 - 2 | 36 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |