Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170749.1 | NP_001164220.1 | 7 - 8 | 1017 | 72 | |
XM_008771008.1 | XP_008769230.1 | 8 - 9 | 1121 | 72 | ||
XM_008771009.1 | XP_008769231.1 | 2 - 3 | 431 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |