Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd40lg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053353.1 | NP_445805.1 | 3 - 4 | 341 | 125 | |
GenBank mRNA | AF116582.1 | - | 3 - 4 | 341 | 125 | |
EF066490.1 | - | 3 - 4 | 341 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |