Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271327.1 | NP_001258256.1 | 1 - 2 | 65 | 82 | |
XM_008773649.1 | XP_008771871.1 | 1 - 2 | 255 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |