Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005533.1 | NP_001005533.1 | 1 - 2 | 88 | 89 | |
XM_006248736.2 | XP_006248798.1 | 1 - 2 | 99 | 89 | ||
GenBank mRNA | BC081998.1 | - | 1 - 2 | 88 | 7131024 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |