Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Card6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106413.1 | NP_001099883.1 | 2 - 3 | 884 | 73 | |
XM_008760760.1 | XP_008758982.1 | 2 - 3 | 1011 | 73 | ||
XM_008760761.1 | XP_008758983.1 | 3 - 4 | 934 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis 3' Most Rattus norvegicus Probe spans exons |