Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107672.1 | NP_001101142.1 | 4 - 5 | 1182 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |