Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017094.1 | NP_058790.1 | 3 - 4 | 329 | 74 | |
XM_008760752.1 | XP_008758974.1 | 3 - 4 | 383 | 74 | ||
XM_008760753.1 | XP_008758975.1 | 3 - 4 | 264 | 74 | ||
XM_008760754.1 | XP_008758976.1 | 3 - 4 | 607 | 74 | ||
GenBank mRNA | BC086591.1 | - | 3 - 4 | 312 | 74 | |
BC103659.1 | - | 3 - 4 | 291 | 74 | ||
BC127501.1 | - | 3 - 4 | 370 | 74 | ||
BC161852.1 | - | 3 - 4 | 316 | 74 | ||
FQ219220.1 | - | 3 - 4 | 361 | 74 | ||
J04811.1 | - | 3 - 4 | 329 | 74 | ||
JF330099.1 | - | 1 - 2 | 69 | 74 | ||
JF412704.1 | - | 2 - 3 | 139 | 74 | ||
S49003.1 | - | 3 - 4 | 329 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |