Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012598.2 | NP_036730.1 | 1 - 2 | 275 | 101 | |
GenBank mRNA | BC081836.1 | - | 1 - 2 | 275 | 7106121 | 101 |
L03294.1 | - | 260 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |