Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025425.1 | NP_001020596.1 | 6 - 7 | 674 | 77 | |
XM_006248768.2 | XP_006248830.1 | 7 - 8 | 859 | 77 | ||
XM_006248769.1 | XP_006248831.1 | 7 - 8 | 864 | 77 | ||
XM_008768985.1 | XP_008767207.1 | 6 - 7 | 815 | 77 | ||
GenBank mRNA | AY879265.1 | - | 6 - 7 | 674 | 77 | |
GU264001.1 | - | 7 - 8 | 722 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |