Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108848.1 | NP_001102318.1 | 4 - 5 | 582 | 101 | |
GenBank mRNA | AB212679.1 | - | 3 - 4 | 430 | 101 | |
BC168972.1 | - | 4 - 5 | 588 | 9034405 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |