Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105903.1 | NP_001099373.1 | 9 - 10 | 1255 | 92 | |
XM_006248754.2 | XP_006248816.1 | 9 - 10 | 1752 | 92 | ||
XM_006248755.2 | XP_006248817.1 | 9 - 10 | 1706 | 92 | ||
XM_006248756.2 | XP_006248818.1 | 9 - 10 | 1750 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |