Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vprbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191798.1 | NP_001178727.1 | 4033 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |