Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106839.1 | NP_001100309.1 | 2 - 3 | 704 | 82 | |
XM_006243405.2 | XP_006243467.1 | 2 - 3 | 1597 | 82 | ||
XM_008766248.1 | XP_008764470.1 | 3 - 4 | 564 | 82 | ||
GenBank mRNA | BC167017.1 | - | 2 - 3 | 596 | 7302522 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |