Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106839.1 | NP_001100309.1 | 1 - 2 | 369 | 99 | |
XM_006243405.2 | XP_006243467.1 | 1 - 2 | 1262 | 99 | ||
GenBank mRNA | BC167017.1 | - | 1 - 2 | 261 | 7302522 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |