Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053965.2 | NP_446417.2 | 3 - 4 | 428 | 67 | |
GenBank mRNA | BC081749.1 | - | 3 - 4 | 428 | 7134670 | 67 |
FQ209681.1 | - | 3 - 4 | 384 | 67 | ||
X97831.1 | - | 3 - 4 | 385 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |