Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108193.1 | NP_001101663.1 | 5 - 6 | 1058 | 110 | |
GenBank mRNA | BC168201.1 | - | 1092 | 7114743 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |