Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112737.2 | NP_001106208.1 | 5 - 6 | 2121 | 76 | |
GenBank mRNA | BC168666.1 | - | 5 - 6 | 2121 | 7933386 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |