Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lztfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024266.1 | NP_001019437.1 | 5 - 6 | 500 | 90 | |
XM_006244191.1 | XP_006244253.1 | 5 - 6 | 644 | 90 | ||
GenBank mRNA | BC078913.1 | - | 5 - 6 | 596 | 90 | |
BC092585.1 | - | 5 - 6 | 500 | 7127181 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |