Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012721.2 | NP_036853.2 | 2 - 3 | 418 | 64 | |
XM_008768851.1 | XP_008767073.1 | 2 - 3 | 608 | 64 | ||
XM_008768852.1 | XP_008767074.1 | 2 - 3 | 293 | 64 | ||
GenBank mRNA | X92070.1 | - | 2 - 3 | 299 | 64 | |
X97376.1 | - | 2 - 3 | 429 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Rattus norvegicus Probe spans exons |