Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc74a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001058720.4 | XP_001058720.2 | 5 - 6 | 2678 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |