Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2n2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021678.1 | NP_067710.1 | 1 | 48 | 85 | |
GenBank mRNA | AF041854.1 | - | 1 | 48 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |