Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105864.1 | NP_001099334.1 | 8 - 9 | 1501 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |