Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100499.1 | NP_001093969.1 | 2270 | 65 | ||
GenBank mRNA | BC092602.1 | - | 6 - 7 | 877 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |