Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100499.1 | NP_001093969.1 | 261 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |