Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100499.1 | NP_001093969.1 | 1754 | 91 | ||
GenBank mRNA | BC092602.1 | - | 3 - 4 | 361 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |