Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fetub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248550.1 | XP_006248612.1 | 1 - 2 | 431 | 90 | |
GenBank mRNA | BC070508.1 | - | 1 - 2 | 53 | 7099154 | 90 |
FQ209449.1 | - | 1 - 2 | 49 | 90 | ||
FQ218557.1 | - | 1 - 2 | 44 | 90 | ||
FQ218884.1 | - | 1 - 2 | 36 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |