Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053837.1 | NP_446289.1 | 5 - 6 | 630 | 65 | |
GenBank mRNA | BC087724.1 | - | 637 | 7320615 | 65 | |
M23674.1 | - | 5 - 6 | 630 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |