Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105867.1 | NP_001099337.1 | 2 - 3 | 480 | 91 | |
XM_006248594.1 | XP_006248656.1 | 2 - 3 | 251 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |