Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013864.1 | NP_001013886.1 | 4 - 5 | 409 | 68 | |
XM_006248505.2 | XP_006248567.1 | 7 - 8 | 846 | 68 | ||
GenBank mRNA | BC083627.1 | - | 4 - 5 | 409 | 7134282 | 68 |
BC083890.1 | - | 3 - 4 | 347 | 7132481 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |