Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768878.1 | XP_008767100.1 | 17 - 18 | 2134 | 77 | |
GenBank mRNA | FQ211899.1 | - | 17 - 18 | 2168 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |