Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108322.1 | NP_001101792.1 | 6 - 7 | 933 | 68 | |
XM_006248619.2 | XP_006248681.1 | 5 - 6 | 1715 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |