Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Proc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012803.1 | NP_036935.1 | 8 - 9 | 847 | 86 | |
XM_006254526.2 | XP_006254588.1 | 8 - 9 | 919 | 86 | ||
XM_006254527.2 | XP_006254589.1 | 8 - 9 | 926 | 86 | ||
XM_008772018.1 | XP_008770240.1 | 8 - 9 | 989 | 86 | ||
GenBank mRNA | BC078879.1 | - | 7 - 8 | 824 | 7122006 | 86 |
X64336.1 | - | 8 - 9 | 847 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |