Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254474.2 | XP_006254536.1 | 3 - 4 | 447 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |