Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108425.1 | NP_001101895.1 | 5 - 6 | 1313 | 111 | |
XM_006254466.2 | XP_006254528.1 | 4 - 5 | 1480 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |