Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108425.1 | NP_001101895.1 | 7 - 8 | 1583 | 68 | |
XM_006254466.2 | XP_006254528.1 | 6 - 7 | 1750 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |