Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751760.3 | XP_003751808.1 | 1 - 2 | 583 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |