Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024373.1 | NP_077349.1 | 8 - 9 | 1404 | 138 | |
XM_006254471.2 | XP_006254533.1 | 9 - 10 | 2825 | 138 | ||
GenBank mRNA | U35890.1 | - | 8 - 9 | 1404 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |