Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017932.1 | NP_001017932.1 | 5 - 6 | 752 | 101 | |
GenBank mRNA | BC127460.1 | - | 5 - 6 | 707 | 8367400 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |